missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcon™ Indicator, For Complexometry, Honeywell Fluka™
Indicator, for complexometry (Al, Fe, Zr)
£28.42 - £63.55
Chemical Identifiers
CAS | 2538-85-4 |
---|---|
Molecular Formula | C20H13N2NaO5S |
Molecular Weight (g/mol) | 416.383 |
MDL Number | MFCD00004069 |
InChI Key | YJUGDMOGPJJWDG-UHFFFAOYSA-M |
PubChem CID | 23702480 |
IUPAC Name | sodium;4-[(2-hydroxynaphthalen-1-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate |
SMILES | C1=CC=C2C(=C1)C=CC(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)[O-])O.[Na+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
2538-85-4 | |
416.383 | |
YJUGDMOGPJJWDG-UHFFFAOYSA-M | |
sodium;4-[(2-hydroxynaphthalen-1-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate |
C20H13N2NaO5S | |
MFCD00004069 | |
23702480 | |
C1=CC=C2C(=C1)C=CC(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)[O-])O.[Na+] |
Specifications
2538-85-4 | |
MFCD00004069 | |
6041435 | |
YJUGDMOGPJJWDG-UHFFFAOYSA-M | |
sodium;4-[(2-hydroxynaphthalen-1-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate | |
23702480 | |
Brown | |
Calcon™ |
C20H13N2NaO5S | |
NONH for all modes of transport | |
Method: 70mg in 10mL N,N-Dimethylformamide | |
C1=CC=C2C(=C1)C=CC(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)[O-])O.[Na+] | |
416.383 | |
416.38g/mol | |
Powder |
Safety and Handling
missing translation for 'rtecsNumber' : 219-810-2
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Calcon
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-1-naphthalenesulphonate