Learn More
Thermo Scientific™ Calibration Validation Carousels
Thermo Scientific™ Performance Verification Standards, Automated (CVC) will ensure the accuracy and reliability of your data while saving time and money.
Brand:  Thermo Scientific™ 10010721
Code : 04
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11556959
  • £5345.00 / Each
Estimated Shipment: 01-08-2024
to see stock.
Description
Description
Thermo Scientific™ Performance Verification Standards, Automated (CVC) will ensure the accuracy and reliability of your data while saving time and money. The CVC provides certified reference materials in a fully automated confirguration for hands-free performance verification of your Thermo Scientific™ spectrophotometer.
  • Simplify qualification procedures by automating performance verification of UV-Visible spectrophotometers
  • Each CVC has a unique serial number matched automatically to the corresponding data files containing standard values and instrument specifications
  • Generated result reports are suitable for printing or archival purposes
  • Included standards are traceable to either NIST™ or NPL primary standards
  • CVCs are available in standard or pharmacopeia compliant configurations
Comprehensive Performance Verification Testing
  • Wavelength Accuracy and Repeatability
  • Photometric Accuracy and Repeatability
  • Stray Light
  • Resolution
  • Photometric Noise
  • Photometric Stability or Drift
  • Baseline Flatness
Save Time, Reduce Errors
  • Eliminates need to copy standard values from certificates
  • Eliminates need to analyze standards individually
  • Eliminates need to match standards to data
  • Eliminates need for manual calculations
  • Eliminates need for manual transcription and reporting of results
Ordering Information:
Unit is shipped in a plastic case for optimal protection during storage. Calibration Validation Carousels can be used with INSIGHT, INSIGHT Security, VISIONpro™, VISIONsecurity™ and applicable Local Control software. Pharmacopeia-compliant CVCs are compatible with the Evolution 600, 300, 260 Bio, 220 and 201 UV-Visible spectrophotometers. Standard configuration CVCs are additionally compatible with the AquaMate Plus, BioMate 6, Evolution 160, Helios Omega, Helios Zeta and UV-10 UV-Visible spectrophotometers.
Specifications
Specifications
All standards contained in the CVC are certified in the Thermo Fisher Scientific ISO/IEC 17025 accredited laboratory traceable to the NIST in the United States or the NPL in the United Kingdom.
Routine Performance Validation - NPL Traceable Filters
Calibration Validation Carousel with Smart Base
NIST traceable
Evolution 201, 220, 260 Bio and 300
10010721