Learn More
Thermo Scientific™ Capit-All™ Screw Cap Tube Capper/Decapper
GREENER_CHOICE
Maintain sample integrity with a hands-free method of capping and decapping tubes quickly and easily.
Brand:  Thermo Scientific™ 4111MAT
40115.92 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10472915
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Maintain sample integrity with a hands-free method of capping and decapping tubes quickly and easily with the Thermo Scientific™ Capit-All™ Screw Cap Tube Capper/Decapper. Designed for benchtop or integrated use, Capit-All instruments cap and decap 24-, 48-, and 96- format Thermo Scientific™ Matrix™ and Thermo Scientific™ Nunc™ ScrewTop tubes with precision and ease. All instruments cap tubes to optimal torque.
Enhanced Productivity
  • Increase throughput by capping and decapping an entire rack at a time in approximately 10 seconds
  • Automatically remove and replace caps on tubes of differing heights, even in the same rack, as well as tubes not properly seated in racks
  • Completely replace manual capping and decapping methods
Superior Sample Protection
  • Caps all tubes in a rack to the optimal torque level, maintaining sample integrity and preventing sample loss during storage
  • Safety features ensure racks are loaded in the correct orientation and that caps are not cross-threaded or overtightened
  • Drip tray prevents cross contamination of samples during addition and removal of tube racks
  • Internal construction ensures debris is not introduced into uncapped tubes
Ease of Use
  • The simple, four-button user interface enables laboratory personnel of any level to operate the instrument
  • Easily integrated into automated systems
  • Easily maintained and able to process up to a million tubes before requiring preventative maintenance
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Matrix Style Screw Top Capped Tubes
50/60 Hz
Capper/Decapper
Capper/Decapper
110 to 240VAC 50/60Hz
45 kg
1
110/240 V
Matrix screw top caps
Approx. 10 Seconds per Rack
96-Channel
14.2 x 12.9 x 22 in. (32.8 x 36.0 x 56.0 cm)
97 lb.
1
Manual via Push Buttons