missing translation for 'onlineSavingsMsg'
Learn More

Erlab™ Captair 632B Smart Filtering Storage Cabinets

These Captair 632B Smart filtering chemical storage cabinets provide outstanding filtration quality to ensure that users and laboratory technicians are protected against chemical hazards. The technology adapts to the type of product being stored and the size is ideal for an underbench position. Users are given the best possible view of the stored products through their transparent doors and are alerted in the event of prolonged opening.

Brand:  Erlab™ AB81//04

Additional Details : Weight : 42.00000kg

 View more versions of this product

Product Code. 17941672

  • £2241.37 / Each
Estimated Shipment: 03-09-2024
to see stock.

Description

Description

  • Air Flow: 11 m3/hr.
  • Structure made of corrosion resistant electro-galvanized steel coated with anti-acid polymer
  • Doors are clear and made with chemical resistant acrylic for easy viewing
  • Filtration module is made with injected polypropylene
  • Carbon filtration: AS for organic vapors, BE for acid vapors, F for formaldehyde vapors, K for ammonia vapors and particulate filtration for powders
  • HEPA H13: 99.95% efficiency filtration of particles over 0.1 μm in size
  • Detection sensor: Type S for solvents, Type A for acids, Type F for formaldehyde
  • Door sensor alarm deploys if doors are left open
  • Key lock
  • Communication Interface: When the light is pulsing: doors left open, containment is compromised, filter breakthrough
  • eGuard APP: For remote control to monitor the status of the storage cabinet, change the settings, and deliver safety alerts immediately to your devices
Specifications

Specifications

28.1 in.
28.1 in.
Steel, Polymer
654 x 583.6 x 715 mm
White
Hinged
2
Chemicals Cabinet
23 in.
1 Tray, 2 Absorbent Mats, 1 Storage Compartment, Storage for 25 x 1 L or 35 x 100 mL or 70 x 50 mL Bottles
25.7 in.
Sliding
1
CE, AFNOR NF X 15-211:2009, EN 1822:1998
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers