missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erlab™ Captair 632B Smart Filtering Storage Cabinets
These Captair 632B Smart filtering chemical storage cabinets provide outstanding filtration quality to ensure that users and laboratory technicians are protected against chemical hazards. The technology adapts to the type of product being stored and the size is ideal for an underbench position. Users are given the best possible view of the stored products through their transparent doors and are alerted in the event of prolonged opening.
£1941.14 - £2241.37
Specifications
Certifications/Compliance | CE, AFNOR NF X 15-211:2009, EN 1822:1998 |
---|---|
Color | White |
Depth (English) Exterior | 25.7 in. |
Dimensions (D x W x H) | 654 x 583.6 x 715 mm |
Height (English) Exterior | 28.1 in. |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
17951672
|
Erlab™
AB81//03 |
2 Trays, 4 Absorbent Mats, 2 Storage Compartments, Storage for 44 x 1 L Bottles |
£1941.14
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17941672
|
Erlab™
AB81//04 |
1 Tray, 2 Absorbent Mats, 1 Storage Compartment, Storage for 25 x 1 L or 35 x 100 mL or 70 x 50 mL Bottles |
£2241.37
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Air Flow: 11 m3/hr.
- Structure made of corrosion resistant electro-galvanized steel coated with anti-acid polymer
- Doors are clear and made with chemical resistant acrylic for easy viewing
- Filtration module is made with injected polypropylene
- Carbon filtration: AS for organic vapors, BE for acid vapors, F for formaldehyde vapors, K for ammonia vapors and particulate filtration for powders
- HEPA H13: 99.95% efficiency filtration of particles over 0.1 μm in size
- Detection sensor: Type S for solvents, Type A for acids, Type F for formaldehyde
- Door sensor alarm deploys if doors are left open
- Key lock
- Communication Interface: When the light is pulsing: doors left open, containment is compromised, filter breakthrough
- eGuard APP: For remote control to monitor the status of the storage cabinet, change the settings, and deliver safety alerts immediately to your devices
Specifications
CE, AFNOR NF X 15-211:2009, EN 1822:1998 | |
25.7 in. | |
28.1 in. | |
Chemicals Cabinet | |
2 |
White | |
654 x 583.6 x 715 mm | |
Steel, Polymer | |
23 in. | |
Sliding |