missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ CaptAvidin™ Agarose (Sedimented Bead Suspension)

CaptAvidin™ Agarose (Sedimented Bead Suspension)

Brand:  Molecular Probes™ C21386

Code : 50

Additional Details : Weight : 0.00525kg

Product Code. 11584237

  • £394.00 / 5mL
Estimated Shipment: 05-08-2024
to see stock.
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

CaptAvidin™ biotin-binding protein conjugated to agarose is designed for use in affinity chromatography. CaptAvidin™ biotin-binding protein contains a nitrated tyrosine in its biotin-binding site. This chemical modification permits dissociation of the avidin-biotin complex under milder conditions than the traditional avidin-biotin complex. At pH 4.0, CaptAvidin™ biotin-binding protein binds biotin tightly with a Ka of ∼109M-1. This association can then be reversed at pH 10, allowing complete dissociation of the avidin-biotin complex. This is in contrast to the harsh conditions (8 M guanidine hydrochloride, pH 1.5) needed to dissociate the avidin-biotin complex.
Specifications

Specifications

Store in refrigerator at 2°C to 8°C
Wet Ice
Agarose
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.