missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Capto™ Core 700
Capto Core 700 is designed for intermediate purification and polishing of viruses and other large biomolecules.
Brand: Cytiva 17-5481-01
This item is not returnable.
View return policy
Description
The novel core bead technology and multimodal, octylamine ligand give Capto Core 700 dual functionality, size exclusion, and binding chromatography in one chromatography medium (resin).
- For intermediate purification and polishing of viruses and other large biomolecules (Mr > 700 000) in flowthrough mode.
- Novel core bead technology allows efficient capture of contaminants while target molecules are collected in the flowthrough.
- Significantly improved productivity and higher flow rates compared with gel filtration methods.
- Straightforward optimization due to flowthrough chromatography and robust performance.
- Convenient small-scale purification, process development, and scale-up using prepacked HiTrap and HiScreen columns.
- Capto Core 700 is composed of a ligand-activated core and inactive shell.
- The inactive shell excludes large molecules (cut off ∼ Mr 700000 [700 kDa]) from entering the core through the pores of the shell.
- These larger molecules are collected in the column flowthrough while smaller impurities bind to the internalized ligands.
- The core of each bead is functionalized with ligands that are both hydrophobic and positively charged, resulting in a highly efficient multimodal binding of various contaminants small enough to enter the core.
- The multimodal ligands ensure strong binding with most impurities over a wide range of pH and salt concentrations.
- Bound impurities are removed from the beads by cleaning-in-place (CIP) procedures using NaOH and in most cases, a solvent.
Specifications
1M NaOH, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol | |
25mL | |
Octylamine | |
90μm | |
4° to 30°C | |
Multimodal Resin |
Capto Core 700 chromato graphy media | |
<500cm/hr. | |
2 to 14 | |
Highly Cross-linked Agarose | |
Intermediate purification and polishing of viruses and other large biomolecules |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Capto Core 700, 25 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.