missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Capture Assay Set
Measurement of cytokines levels in-vivo
Brand: BD 558491
Additional Details : Weight : 0.50000kg
Includes: Plate coating antibody, in vivo capture antibody, IVC standard, enzyme concentrate, technical data sheet and lot-specific certificate of analysis
Description
- Reactivity: Mouse
- Format: Set
In-vitro capture assay
Specifications
Plate coating Antibody, in vivo capture Antibody, IVC standard, Enzyme concentrate, technical data sheet and lot specific certificate of analysis | |
Mouse IFN-gamma capture assay set | |
10 Plates | |
Set |
Research Use Only