missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Capture Assay Set
Measurement of cytokines levels in-vivo
£932.52
Specifications
Includes | Plate coating Antibody, in vivo capture Antibody, IVC standard, Enzyme concentrate, technical data sheet and lot specific certificate of analysis |
---|---|
Quantity | 10 Plates |
Format | Set |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
12850470
|
BD
558491 |
Mouse IFN-gamma capture assay set |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Reactivity: Mouse
- Format: Set
In-vitro capture assay
Specifications
Plate coating Antibody, in vivo capture Antibody, IVC standard, Enzyme concentrate, technical data sheet and lot specific certificate of analysis | |
10 Plates | |
Set |
Research Use Only