missing translation for 'onlineSavingsMsg'
Learn More
Carbon, activated, -20+40 mesh, Thermo Scientific Chemicals Product Code.: 15442297

Carbon, activated, -20+40 mesh, Thermo Scientific Chemicals

Product Code. 15442297
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 15442297

Brand: Thermo Scientific Alfa Aesar 045478.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
Mesh Size -20+40 Mesh
Quantity 100 g
Physical Form Powder
Chemical Name or Material Carbon

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP31I026 Certificate TypeCertificate of Analysis Date19/08/2022 Catalog Number045480.AA,045478.22,045478.36,045478.A3
Lot NumberS27A011 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number045477.30,045478.22,045477.18
Lot NumberZ12D013 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number045477.30,045478.22,045477.18
Lot Number10194109 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number045477.30,045478.22,045477.18
Lot NumberG12W049 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number045477.30,045478.22,045477.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Carbon, activated, -20+40 mesh, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.