missing translation for 'onlineSavingsMsg'
Learn More

Carbon slug, activated, 6-8 mesh, Thermo Scientific Chemicals

For Hydrocarbon Recovery

£59.30 - £162.81

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11329349
View Documents Promotion Details
Thermo Scientific Alfa Aesar
088764.A1
1 kg
£59.30
1kg
Please to purchase this item. Need a web account? Register with us today!
11319349
View Documents
Thermo Scientific Alfa Aesar
088764.A7
5 kg
£162.81
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For hydrocarbon recovery

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Activity 50% min.
Specifications

Chemical Identifiers

7440-44-0
12.011
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00133992
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]

Specifications

7440-44-0
6-8. Mesh
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
Coarse Powder, Granular, Pellets or Slugs
Black
C
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
[C]
12.011
CHEBI:27594
Carbon
Videos
Safety and Handling

Safety and Handling

H500

missing translation for 'einecsNumber' : 231-153-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only