Learn More
Swann-Morton™ Carbon Steel Non-sterile Scalpel Blades
Use Swann Morton™ Carbon Steel Non-sterile Scalpel Blades for precise cutting in the operating room.
Brand: Swann-Morton™ 0112
This item is not returnable.
View return policy
Description
Use Swann Morton™ Carbon Steel Non-sterile Scalpel Blades for precise cutting in the operating room.
- Carbon steel to BS EN 27740
- Handles and blades sold separately
Specifications
Carbon Steel | |
# 25 | |
10/Pk. |
Scalpel Blade | |
Non-sterile |
Product Content Correction
Product Title
Swann-Morton™ Carbon Steel Non-sterile Scalpel Blades > Size: No. 25
Spot an opportunity for improvement?Share a Content Correction