Learn More
Swann-Morton™ Carbon Steel Sterile Scalpel Blades
Use for convenience in user environments: on the ward, in emergency or trauma units or even out in the field with paramedics. Swann Morton™ Carbon Steel Surgical Scalpel Blades come in single peel packs.
Brand: Swann-Morton™ 0203
This item is not returnable.
View return policy
Description
- Carbon steel to BS EN 27740
- Sterile
Specifications
Carbon Steel | |
# 11 | |
Sterile | |
10/Pk. |
Scalpel Blade | |
The # 11 Blade fit Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L | |
CE compliant 93/42/EC |