Learn More
Swann-Morton™ Carbon Steel Sterile Scalpel Blades
Use for convenience in user environments: on the ward, in emergency or trauma units or even out in the field with paramedics. Swann Morton™ Carbon Steel Surgical Scalpel Blades come in single peel packs.
Brand:  Swann-Morton™ 0210
 View more versions of this product
Product Code. 11798363
  • £22.80 / Pack of 100
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Carbon steel to BS EN 27740
  • Sterile
Specifications
Specifications
Carbon Steel
# 23
Sterile
10/Pk.
Scalpel Blade
The # 23 fits Handles # 4, # 4L, # 4 Graduated and # 6B
CE compliant 93/42/EC