missing translation for 'onlineSavingsMsg'
Learn More

Carbonic Anhydrase I Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA597526

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16500362

  • £386.00 / 100µL
Estimated Shipment: 24-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Carbonic anhydrases (CAs) are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. CA1 is closely linked to CA2 and CA3 genes on chromosome 8, and it encodes a cytosolic protein which is found at the highest level in erythrocytes. Variants of this gene have been described in some populations. Multiple alternatively spliced variants, encoding the same protein, have been identified. Transcript variants of CA1 utilizing alternative polyA_sites have been described in literature.
Specifications

Specifications

Carbonic Anhydrase I
Polyclonal
Unconjugated
CA1
AW555628, CA I, CA1, ca1 {ECO:0000250, CAB, CA-I, CA-I {ECO:0000250, Car1, Car-1, Carbonate dehydratase I, carbonate dehydratase I {ECO:0000250, carbonic anhydrase 1, carbonic anhydrase 1 {ECO:0000250, Carbonic anhydrase B, carbonic anhydrase I, carbonic anhydrase I {ECO:0000250, epididymis secretory protein Li 11, HEL-S-11, UniProtKB:P00915}
Rabbit
Antigen affinity chromatography
RUO
12346, 759
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
P00915, P13634
CA1, Car1
Recombinant Human Carbonic anhydrase 1 protein (1-261AA)
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers