missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase I Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
£386.00 - £387.00
Specifications
Antigen | Carbonic Anhydrase I |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16510362
|
Invitrogen
PA597527 |
100 μg |
£387.00
100µg |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16500362
|
Invitrogen
PA597526 |
100 μL |
£386.00
100µL |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Carbonic anhydrases (CAs) are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. CA1 is closely linked to CA2 and CA3 genes on chromosome 8, and it encodes a cytosolic protein which is found at the highest level in erythrocytes. Variants of this gene have been described in some populations. Multiple alternatively spliced variants, encoding the same protein, have been identified. Transcript variants of CA1 utilizing alternative polyA_sites have been described in literature.Specifications
Carbonic Anhydrase I | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
AW555628, CA I, CA1, ca1 {ECO:0000250, CAB, CA-I, CA-I {ECO:0000250, Car1, Car-1, Carbonate dehydratase I, carbonate dehydratase I {ECO:0000250, carbonic anhydrase 1, carbonic anhydrase 1 {ECO:0000250, Carbonic anhydrase B, carbonic anhydrase I, carbonic anhydrase I {ECO:0000250, epididymis secretory protein Li 11, HEL-S-11, UniProtKB:P00915} | |
CA1, Car1 | |
Primary | |
Antibody |
Polyclonal | |
Liquid | |
RUO | |
P00915, P13634 | |
12346, 759 | |
IgG | |
-20°C or -80°C if preferred | |
CA1 |