missing translation for 'onlineSavingsMsg'
Learn More

Carbonic Anhydrase VA Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

£146.00 - £337.00

Specifications

Antigen Carbonic Anhydrase VA
Concentration 0.5 mg/mL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17948792
View Documents
Invitrogen
PA5145354
20 μL
£146.00
20µL
Please to purchase this item. Need a web account? Register with us today!
17989522
View Documents
Invitrogen
PA5145353
200 μL
£337.00
200µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric juice. They show extensive diversity in distribution and in their subcellular localization. The human CA5A gene encodes for the CA V protein which is expressed in the mitochondrial matrix in liver tissues, but can also be detected in many other tissues. In ureagenesis, the CA V protein may be involved in providing bicarbonate to carbamoyl phosphate synthetase.
Specifications

Specifications

Carbonic Anhydrase VA
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Mouse, Rat
P23589, P43165
12352, 54233
Recombinant Rat CA5A protein, Tyr12-Leu219 (Accession #P43165)
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4
CA Y, CA5, CA5A, CA5AD, Car5, Car5a, Carbonate dehydratase VA, carbonic anhydrase 5 mitochondrial, carbonic anhydrase 5, mitochondrial, carbonic anhydrase 5A, carbonic anhydrase 5a, mitochondrial, carbonic anhydrase 5b, mitochondrial, carbonic anhydrase V, mitochondrial, carbonic anhydrase VA, carbonic anhydrase VA, mitochondrial, carbonic dehydratase, CAV, CAVA, CA-VA, GS1-21A4.1
CA5A, Car5a
IgG
Protein A
Car5a
Videos
SDS
Documents

Documents

Product Certifications