missing translation for 'onlineSavingsMsg'
Learn More

Carbonic Anhydrase VIII Monoclonal Antibody (1G5G12), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA542990

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 17759543

  • £350.00 / 100µL
Estimated Shipment: 02-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specificity: Human Carbonic Anhydrase VIII/CA8

The protein encoded by this gene was initially named CA-related protein because of sequence similarity to other known carbonic anhydrase genes. However, the gene product lacks carbonic anhydrase activity (i.e., the reversible hydration of carbon dioxide). The gene product continues to carry a carbonic anhydrase designation based on clear sequence identity to other members of the carbonic anhydrase gene family. The absence of CA8 gene transcription in the cerebellum of the lurcher mutant in mice with a neurologic defect suggests an important role for this acatalytic form.
Specifications

Specifications

Carbonic Anhydrase VIII
Monoclonal
1 mg/mL
PBS with no preservative
P35219
Ca8
Recombinant Human Carbonic Anhydrase VIII/CA8 protein.
100 μL
Primary
Human
Antibody
IgG2a
ELISA
1G5G12
Unconjugated
Ca8
AW546993; Ca8; Cals; Cals1; CAMRQ3; Car8; carbonate dehydratase; carbonic anhydrase 8; carbonic anhydrase VIII; carbonic anhydrase-like sequence; carbonic anhydrase-like sequence 1; carbonic anhydrase-related protein; CA-related protein; CARP; CA-RP; CARP VIII; CA-RP VIII; CA-VIII; hypothetical protein LOC550233; LOW QUALITY PROTEIN: carbonic anhydrase-related protein; MGC120502; MGC99509; wdl; zgc:110118
Mouse
Protein A
RUO
767
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers