missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase VIII Monoclonal Antibody (1G5G12), Invitrogen™
Mouse Monoclonal Antibody
£199.00 - £350.00
Specifications
Antigen | Carbonic Anhydrase VIII |
---|---|
Clone | 1G5G12 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17789853
|
Invitrogen
MA543510 |
50 μL |
£199.00
50µL |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17759543
|
Invitrogen
MA542990 |
100 μL |
£350.00
100µL |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Specificity: Human Carbonic Anhydrase VIII/CA8
The protein encoded by this gene was initially named CA-related protein because of sequence similarity to other known carbonic anhydrase genes. However, the gene product lacks carbonic anhydrase activity (i.e., the reversible hydration of carbon dioxide). The gene product continues to carry a carbonic anhydrase designation based on clear sequence identity to other members of the carbonic anhydrase gene family. The absence of CA8 gene transcription in the cerebellum of the lurcher mutant in mice with a neurologic defect suggests an important role for this acatalytic form.Specifications
Carbonic Anhydrase VIII | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
AW546993; Ca8; Cals; Cals1; CAMRQ3; Car8; carbonate dehydratase; carbonic anhydrase 8; carbonic anhydrase VIII; carbonic anhydrase-like sequence; carbonic anhydrase-like sequence 1; carbonic anhydrase-related protein; CA-related protein; CARP; CA-RP; CARP VIII; CA-RP VIII; CA-VIII; hypothetical protein LOC550233; LOW QUALITY PROTEIN: carbonic anhydrase-related protein; MGC120502; MGC99509; wdl; zgc:110118 | |
Ca8 | |
IgG2a | |
Protein A | |
Antibody |
1G5G12 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P35219 | |
767 | |
Recombinant Human Carbonic Anhydrase VIII/CA8 protein. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Ca8 |