Learn More
Cytiva Cassette
For use with the Fraction collector F9-C and the ÄKTA avant chromatography system. Cytiva Cassettes are available for Deep-Well Plates or 3mL Tubes for large-scale processing of samples in chromatography applications. Pack of 2.
Brand:  Cytiva 28954212
 View more versions of this product
Product Code. 12229319
  • £141.00 / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Cytiva Cassettes are available in packs of two (2) for large-scale processing of samples in chromatography applications using the Fraction collector F9-C and the ÄKTA avant chromatography system.
  • For Deep-Well Plates (in Flexible fraction collector): Holds 24, 48 or 96 deep-well plate
  • For 3mL Tubes: Holds up to 40 tubes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chromatography Applications
Cassette
For Use With 24, 48 or 96 deep-well plate
Fraction Collector F9-C and ÄKTA Avant Chromatography System
2/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Cassette > For Use With: 24, 48 or 96 deep-well plate