missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cassette Foam Sponges

These Cassette Foam Sponges are accessory items for transfer tanks.

Brand:  Cytiva TE25F-1/8

Code : GE

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 10279394

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Cassette foam sponges
Specifications

Specifications

TE 22 Mini Tank Transfer Unit
Foam
0.11 in.
Cassette Foam Sponge
4 Pk.
3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers