missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Casting Base

Accessory for use with Vertical Gel Electrophoresis System.

Brand:  Fisherbrand™ VS10DCAST

Product Code. 11847623

  • £104.00 / Each
Estimated Shipment: 24-06-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Measures 100 x 100mm
Specifications

Specifications

Vertical Gel Electrophoresis System
Casting Base, 100 x 100 mm
Product Suggestions

Product Suggestions

Videos

Videos

  • Fisherbrand and Fisher BioReagents Focus on electrophoresis

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Casting Base >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.