missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catalase Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1F8D7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66765-1-IG-150UL
This item is not returnable.
View return policy
Description
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kD in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara′s disease.Specifications
Catalase | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O62839, P04040, P04762, P24270 | |
CAT | |
Catalase Fusion Protein Ag16916 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG1 |
Western Blot | |
1F8D7 | |
Unconjugated | |
CAT | |
CAT, catalase | |
Mouse | |
Protein G | |
RUO | |
12359, 24248, 397568, 847 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction