missing translation for 'onlineSavingsMsg'
Learn More

Catalase Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1F8D7, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen Catalase
Clone 1F8D7
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16828945
View Documents
Proteintech
66765-1-IG-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16818945
View Documents
Proteintech
66765-1-IG-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kD in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara′s disease.
Specifications

Specifications

Catalase
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CAT, catalase
CAT
IgG1
Protein G
Antibody
1F8D7
Western Blot
Unconjugated
Mouse
Human, Mouse, Rat, Porcine
O62839, P04040, P04762, P24270
12359, 24248, 397568, 847
Catalase Fusion Protein Ag16916
Primary
-20°C
CAT
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.