Learn More
Cole-Parmer™ Catalyst Pump Tubing
Designed especially for handling food products. Cole-Parmer™ Catalyst Tygon E-Food Pump Tubing is compatible with Catalyst FH100, FH100D Pumps.
Brand:  Cole-Parmer™ MFLX97000-72
 View more versions of this product
Product Code. 15435613
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Designed for use with Catalyst FH100, FH100D, FH100X and FH100DX Pumps. Cole-Parmer™ Catalyst Pump Tubing is available four formulations in 15 meter lengths.
  • Transparent pump tubing
  • For Catalyst FH100, FH100D, FH100X and FH100DX Pumps
  • Four formulations available to cover most chemical compatible applications
  • To help simplify choices, a platinum-cured silicone, two TPEs and a long life PVC tubing material are offered.
  • Standard 15 meter lengths
  • Available in both thin and thick wall configurations
  • FH100 and FH100D utilize tubing with a wall thickness of 1.6mm
  • FH100X and FH100DX utilize tubing with a wall thickness of 2.4mm
Specifications
Specifications
Catalyst Pump Coiled Tubing
3.2 mm
49.2 ft.
0.094 in.
Catalyst Pump models: FH100, FH100D, FH100X and FH100DX
0.12 in.
Silicone (Platinum-Cured)
1500 cm
1.6 mm
Clear