Learn More
Catechol, 99+%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Melting Point | 103.0°C to 106.0°C |
Color | Brown-Gray to White |
Boiling Point | 245.0°C |
Flash Point | 131°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | Glass bottle |
Quantity | 100 g |
Merck Index | 15,8111 |
Solubility Information | Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol, benzene, chloroform, ether, and pyridine and aqueous solutions |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0463594 | Certificate of Analysis | 12/06/2024 | 158980100,158980025,158981000,158985000,158980050 |
A0420578 | Certificate of Analysis | 05/01/2023 | 158980100,158980025,158981000,158985000,158980050 |
A0403242 | Certificate of Analysis | 15/12/2022 | 158980100,158980025,158981000,158985000,158980050 |
A0401689 | Certificate of Analysis | 15/12/2022 | 158980100,158980025,158981000,158985000,158980050 |
A0402605 | Certificate of Analysis | 14/12/2022 | 158980100,158980025,158981000,158985000,158980050 |
Safety and Handling



- Catechol
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H341-Suspected of causing genetic defects.
- H311-Toxic in contact with skin.
- H332-Harmful if inhaled.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.