missing translation for 'onlineSavingsMsg'
Learn More

Catechol, 99+%, Thermo Scientific Chemicals

£16.18 - £495.47

Chemical Identifiers

CAS 120-80-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00002188
InChI Key YCIMNLLNPGFGHC-UHFFFAOYSA-N
Synonym pyrocatechol, catechol, 1,2-dihydroxybenzene, 1,2-benzenediol, pyrocatechin, 2-hydroxyphenol, o-benzenediol, pyrocatechine, o-dihydroxybenzene, o-dioxybenzene
PubChem CID 289
ChEBI CHEBI:18135
IUPAC Name benzene-1,2-diol
SMILES OC1=CC=CC=C1O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10051720
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Acros
158980050
5 g Glass bottle
£17.99
5g
Please to purchase this item. Need a web account? Register with us today!
10756581
missing translation for 'viewDocuments'
Thermo Scientific Acros
158981000
100 g Glass bottle
£16.18
100g
Please to purchase this item. Need a web account? Register with us today!
10337860
missing translation for 'viewDocuments'
Thermo Scientific Acros
158985000
500 g Glass bottle
£36.88
500g
Please to purchase this item. Need a web account? Register with us today!
10542354
missing translation for 'viewDocuments'
Thermo Scientific Acros
158980025
2.5 kg Plastic Bottle
£127.64
2.50kg
Please to purchase this item. Need a web account? Register with us today!
11954671
missing translation for 'viewDocuments'
Thermo Scientific Acros
158980100
10 kg Fibre drum
£495.47
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

120-80-9
110.11
YCIMNLLNPGFGHC-UHFFFAOYSA-N
289
benzene-1,2-diol
C6H6O2
MFCD00002188
pyrocatechol, catechol, 1,2-dihydroxybenzene, 1,2-benzenediol, pyrocatechin, 2-hydroxyphenol, o-benzenediol, pyrocatechine, o-dihydroxybenzene, o-dioxybenzene
CHEBI:18135
OC1=CC=CC=C1O

Specifications

120-80-9
Brown-Gray to White
131°C
99% min. (GC)
MFCD00002188
pyrocatechol, catechol, 1,2-dihydroxybenzene, 1,2-benzenediol, pyrocatechin, 2-hydroxyphenol, o-benzenediol, pyrocatechine, o-dihydroxybenzene, o-dioxybenzene
YCIMNLLNPGFGHC-UHFFFAOYSA-N
benzene-1,2-diol
289
2.38 mm2/s (110°C)
99+%
Catechol
103.0°C to 106.0°C
245.0°C
Authentic
C6H6O2
15,8111
Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol, benzene, chloroform, ether, and pyridine and aqueous solutions
OC1=CC=CC=C1O
110.11
CHEBI:18135
110.11
Crystalline Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing genetic defects.
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.<b

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cau

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 204-427-5

Recommended Storage : Product may darken during storage

Product Identifier
  • Catechol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H311-Toxic in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.