Learn More
Tocris Bioscience™ CB 65
High affinity, selective CB2 agonist
Brand:  Tocris Bioscience™ 2663/50
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18075042
  • £734.83 / 50mg
Estimated Shipment: 27-08-2024
to see stock.
Description
Description
CB 65 is a high affinity and selective CB2 receptor agonist; Ki values are 3.3 and > 1000 nM for CB2 and CB1 receptors respectively.
Specifications
Specifications
N-Cyclohexyl-7-chloro-1-[2-(4-morpholinyl)ethyl]quinolin-4(1H)-one-3-carboxamide
50 mg
C22H28ClN3O3
913534-05-1
Cannabinoid R2/CB2 Agonists
0.99