missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CB 65
High affinity, selective CB2 agonist
£209.16 - £734.83
Specifications
Chemical Name or Material | N-Cyclohexyl-7-chloro-1-[2-(4-morpholinyl)ethyl]quinolin-4(1H)-one-3-carboxamide |
---|---|
CAS | 913534-05-1 |
Target | Cannabinoid R2/CB2 Agonists |
Molecular Formula | C22H28ClN3O3 |
Purity | 0.99 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18025182
|
Tocris Bioscience™
2663/10 |
10 mg |
£209.16
10mg |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18075042
|
Tocris Bioscience™
2663/50 |
50 mg |
£734.83
50mg |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CB 65 is a high affinity and selective CB2 receptor agonist; Ki values are 3.3 and > 1000 nM for CB2 and CB1 receptors respectively.Specifications
N-Cyclohexyl-7-chloro-1-[2-(4-morpholinyl)ethyl]quinolin-4(1H)-one-3-carboxamide | |
Cannabinoid R2/CB2 Agonists | |
0.99 |
913534-05-1 | |
C22H28ClN3O3 |