missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tango™ CCR5-bla U2OS Cells
Tango™ CCR5-bla U2OS Cells contain the human Chemokine (C-C Motif) Receptor 5 (CCR5) linked to a TEV protease site.
Brand: Tango™ K1788
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Gene Symbol: CCR5
- Accession Number: NM_001295
- Cytoplasmic Tail: Native Tail
- Population Status: Single Cell Clone Origin
- The Tango™ CCR5-bla U2OS cells contain the human Chemokine (C-C Motif) Receptor 5 (CCR5) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
- The Tango™ CCR5-bla U2OS cells are functionally validated for Z' and EC50 concentrations of MIP-1a. In addition, Tango™ CCR5-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
Specifications
Liquid Nitrogen | |
GPCRs | |
8 x 106 Cells |
Fluorescent | |
CCR5-bla U2OS cells, CCR5-bla U2OS Assay Protocol, Certificate of Analysis | |
Dividing Cells |