missing translation for 'onlineSavingsMsg'
Learn More

CD18 Mouse anti-Human, FITC, Clone: 6.7, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 555923

166.17 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15868528

  • £169.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The 6.7 monoclonal antibody specifically binds to β2 integrin, a 90-95 kDa transmembrane protein, which associates non-covalently as a heterodimer with the a chains of CD11a, CD11b, or CD11c. CD18 is expressed on lymphocytes, monocytes, and more weakly on granulocytes. LFA-1 has been shown to play an important role in homotypic and heterotypic cellular adhesion in immune and inflammatory responses.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD18
Monoclonal
FITC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Cloned Human DS6 TCR γδ+ Cells
100 Tests
Immunology
Cynomolgus, Human, Pig, Rabbit, Rhesus Monkey
IgG1 κ
Flow Cytometry
6.7
ITGB2; Integrin beta-2; Beta subunit for LFA-1, MAC-1/CR3, and p150,95
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD18 Mouse anti-Human, FITC, Clone: 6.7, BD > 100 Tests; FITC

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.