missing translation for 'onlineSavingsMsg'
Learn More

CD18 Mouse anti-Human, FITC, Clone: 6.7, BD

Mouse Monoclonal Antibody

£169.00

Specifications

Antigen CD18
Clone 6.7
Applications Flow Cytometry
Classification Monoclonal
Conjugate FITC
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15868528
View Documents
BD Biosciences
555923
100 Tests
£169.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 6.7 monoclonal antibody specifically binds to β2 integrin, a 90-95 kDa transmembrane protein, which associates non-covalently as a heterodimer with the a chains of CD11a, CD11b, or CD11c. CD18 is expressed on lymphocytes, monocytes, and more weakly on granulocytes. LFA-1 has been shown to play an important role in homotypic and heterotypic cellular adhesion in immune and inflammatory responses.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

CD18
Flow Cytometry
FITC
RUO
Cynomolgus, Human, Pig, Rabbit, Rhesus Monkey
Cloned Human DS6 TCR γδ+ Cells
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
6.7
Monoclonal
Mouse
Immunology
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
ITGB2; Integrin beta-2; Beta subunit for LFA-1, MAC-1/CR3, and p150,95
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD18 Mouse anti-Human, FITC, Clone: 6.7, BD

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.