Learn More
CD22 Mouse anti-Human, CoraLite 488, Clone: 7A2F1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66103-100UL
Product Code. 16820726
  • £375.00 / 100µL
Explore more special offers
Description
Description
The CD22 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human CD22. This antibody recognizes human antigen. The CD22 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
CD22
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Immunofluorescence
7A2F1
CoraLite 488
100 μL
933
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
CD22 Mouse anti-Human, CoraLite 488, Clone: 7A2F1, Proteintech > 100 μL