missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD23 Mouse, PE, Clone: EBVCS-5, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB32-PE-25Tests
This item is not returnable.
View return policy
Specifications
CD23 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
FCER2 | |
Protein A/G Purified | |
RUO | |
2208 | |
IgG1 κ |
Flow Cytometry | |
EBVCS-5 | |
PE | |
NP_001993 | |
Mouse | |
25 Tests | |
Primary | |
Store at 2-4°C. |