missing translation for 'onlineSavingsMsg'
Learn More

CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH5), NovaFluor™ Red 755, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

£132.00 - £256.00

Specifications

Antigen CD274 (PD-L1, B7-H1)
Clone MIH5
Concentration 0.1 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17806872
View Documents
Invitrogen
M036T02R06
25 μg
£132.00
25µg
Please to purchase this item. Need a web account? Register with us today!
17857372
View Documents
Invitrogen
M036T03R06
100 μg
£256.00
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.
Specifications

Specifications

CD274 (PD-L1, B7-H1)
0.1 μg/Test
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1
Cd274
Primary
4° C, store in dark, DO NOT FREEZE!
MIH5
Flow Cytometry
NovaFluor Red 755
Rat
Mouse
Q9EP73
60533
IgG2a λ
Affinity chromatography
Cd274
Videos
SDS
Documents

Documents

Product Certifications