Learn More
IBA Lifesciences CD4/CD62L/CD45RA Fab-TACS™ Magnetic Microbead Starter Kit
Label-free isolation of central memory T cells from PBMCs
Brand: IBA Lifesciences 6-8000-115
This item is not returnable.
View return policy
Description
- Fab-TACS™ - Traceless Affinity Cell Selection - is a positive cell isolation approach based on our Strep-tag™ technology CD4/CD62L/CD45RA Fab-TACS™ Magnetic Microbead Starter Kit allows label-free isolation of CD4+ central memory T cells (CD4+CD62L+CD45RA-) from peripheral blood mononuclear cells (PBMCs)
- Alternatively, CD4/CD62L/CD45RO Fab-TACS™ Magnetic Microbead Starter Kit is suitable for the isolation of these cells.
- Low affinity, Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) are incubated with Strep-Tactin™ Magnetic Microbeads to form a complex that stably binds to the cell surface. After specific labeling, target cells are bound to a magnet and non-target cells efficiently washed away.
- In a final step, the addition of biotin causes the release of the isolated cells from magnetic microbeads. The Fab-Streps self-dissociate from the cell surface due to their low affinity.
- After removing biotin by washing, the isolated cells are ready for further positive selection or depletion steps or can directly be used for further downstream applications
Specifications
Strep-Tactin™ Magnetic Microbeads, CD4, CD62L, CD45RA Fab-Strep (human), 10 x Buffer CI, Biotin stock solution | |
Label-Free Isolation of Central Memory T Cells From PBMCs |