missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD4 Fab-Strep
Low affinity Fab fragment fused to a Twin-Strep-tag™
£24.25
Specifications
Form | Lyophilized |
---|---|
For Use With (Application) | Fluorescent Cell Staining & Sorting, Magnetic or Affinity Chromatographic Cell Isolation |
Description
- CD4 Fab-Strep is a recombinantly expressed low affinity Fab fragment fused to a Twin-Strep-tag™
- Due to its low affinity for the target, an increase in avidity is required to ensure sufficient binding to the cell surface
- Combining Fab-Streps with a Strep-Tactin™ backbone leads to the formation of complexes that stably bind to cells
- Fab-Streps were developed for Traceless Affinity Cell Selection (Fab-TACS™), which is based on our Strep-tag™ technology
Specifications
Lyophilized | |
Fluorescent Cell Staining & Sorting, Magnetic or Affinity Chromatographic Cell Isolation |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title