missing translation for 'onlineSavingsMsg'
Learn More

CD40L/CD154 Mouse anti-Human, Clone: 1E6D10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66502-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888435

  • £130.00 / 20µL
Estimated Shipment: 21-08-2024
to see stock.

Description

Description

The CD40 antigen is a single chain glycoprotein. It is known to be a member of the tumor necrosis factor / nerve growth factor superfamily and shows a significant homology to the Hodgkin′s disease associated antigen, CD30. CD40 is present on all B cells except plasma cells. It is also found on some epithelial cells, carcinomas and lymphoid dendritic cells. The precise function of the CD40 antigen is unknown but it appears to be involved in the transduction of regulatory signals for cellular functions such as B cell proliferation and differentiation.
Specifications

Specifications

CD40L/CD154
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P29965
CD40LG
CD40L/CD154 Fusion Protein Ag24941
20 μL
Primary
Human
Antibody
IgG2a
Western Blot, Immunohistochemistry (Paraffin)
1E6D10
Unconjugated
CD40LG
CD154, CD40 L, CD40 ligand, CD40L, CD40LG, gp39, hCD40L, HIGM1, IGM, IMD3, T BAM, T cell antigen Gp39, TNF related activation protein, TNFSF5, TRAP
Mouse
Protein A
RUO
959
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers