missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40L/CD154 Mouse anti-Human, Clone: 1E6D10, Proteintech
Mouse Monoclonal Antibody
£130.00 - £339.00
Specifications
Antigen | CD40L/CD154 |
---|---|
Clone | 1E6D10 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888435
|
Proteintech
66502-1-IG-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16878435
|
Proteintech
66502-1-IG-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The CD40 antigen is a single chain glycoprotein. It is known to be a member of the tumor necrosis factor / nerve growth factor superfamily and shows a significant homology to the Hodgkin′s disease associated antigen, CD30. CD40 is present on all B cells except plasma cells. It is also found on some epithelial cells, carcinomas and lymphoid dendritic cells. The precise function of the CD40 antigen is unknown but it appears to be involved in the transduction of regulatory signals for cellular functions such as B cell proliferation and differentiation.Specifications
CD40L/CD154 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CD154, CD40 L, CD40 ligand, CD40L, CD40LG, gp39, hCD40L, HIGM1, IGM, IMD3, T BAM, T cell antigen Gp39, TNF related activation protein, TNFSF5, TRAP | |
CD40LG | |
IgG2a | |
Protein A | |
Antibody |
1E6D10 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P29965 | |
959 | |
CD40L/CD154 Fusion Protein Ag24941 | |
Primary | |
-20°C | |
CD40LG |