missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDON Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | CDON |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16837574
|
Proteintech
17634-1-AP-20UL |
20 μL |
£130.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16827574
|
Proteintech
17634-1-AP-150UL |
150 μL |
£339.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDON and BOC are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III (FNIII. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.Specifications
CDON | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O35158, Q32MD9, Q4KMG0 | |
50937, 50938, 57810 | |
CDON Fusion Protein Ag11904 | |
Primary | |
-20°C | |
CDON |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CDO, CDON, Cdon homolog (mouse), ORCAM | |
CDON | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |