Learn More
CDON Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17634-1-AP-20UL
 View more versions of this product
Product Code. 16837574
  • £135.00 / 20µL
Explore more special offers
Description
Description
CDON and BOC are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III (FNIII. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.
Specifications
Specifications
CDON
Polyclonal
Unconjugated
CDON
CDO, CDON, Cdon homolog (mouse), ORCAM
Rabbit
Antigen Affinity Chromatography
RUO
50937, 50938, 57810
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35158, Q32MD9, Q4KMG0
CDON
CDON Fusion Protein Ag11904
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
CDON Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated