Learn More
Molecular Probes™ CDP-Star™ Substrate (0.25 mM Ready-To-Use)
The CDP-Star™ Substrate is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes.
Brand:  Molecular Probes™ T2145
 View more versions of this product
Product Code. 10125425
  • £209.00 / 50mL
Estimated Shipment: 11-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The CDP-Star™ Substrate is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.
This CDP-Star™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.
  • Chemiluminescent substrate provides highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
  • Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.
This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDP-Star™ Substrate, 50mL, Liquid Form, Chemiluminescent Alkaline Phosphatase Substrate, 0.25mM Ready-to-use For Protein or Nucleic Acid Blotting on Nitrocellulose membranes, Protein Assays and Analysis, Southern, Northern and Western blotting, Can also be used in Solution-based Assays
50 mL
Liquid
Protein assays and analysis, southern, northern, western blotting, can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays, protein biology, cell analysis, enzyme reporter assays, ELISA, alkaline phosphatase assays, build your own immunoassay, ready-to-use immunoassay
Alkaline Phosphatase
Room Temperature
ELISA Substrate
Chemiluminescence
Bottle
CDP-Star™, NovaBright™
Store the product at 4°C.