Learn More
Molecular Probes™ CDP-Star™ Substrate (0.25 mM Ready-To-Use)
The CDP-Star™ Substrate is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes.
£209.00 - £608.00
Specifications
Product Type | ELISA Substrate |
---|---|
Form | Liquid |
Format | Bottle |
Product Line | CDP-Star™, NovaBright™ |
Substrate Type | Alkaline Phosphatase |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10125425
|
Molecular Probes™
T2145 |
50 mL |
£209.00
50mL |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
10144664
|
Molecular Probes™
T2146 |
100 mL |
£243.00
100mL |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||||
12674325
|
Molecular Probes™
T2147 |
250 mL |
£608.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
Description
The CDP-Star™ Substrate is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.
This CDP-Star™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.
- Chemiluminescent substrate provides highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
- Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.
This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours.
Specifications
ELISA Substrate | |
Bottle | |
Alkaline Phosphatase |
Liquid | |
CDP-Star™, NovaBright™ | |
Store the product at 4°C. |
For Research Use Only. Not for use in diagnostic procedures.