Learn More
Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassays, Calibrators and Controls
Complete your ISD panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay.
Brand: Thermo Scientific™ 100278
Description
Complete your immunosuppressant drug testing panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay. Like our other ISD assays, the CEDIA Mycophenolic Acid Immunoassay is accurate and precise, providing users with confidence in their results. Applications are available on our flexible and intuitive Indiko™ and Indiko Plus clinical and specialty chemical systems as well as a wide variety of other laboratory instrumentation.
Accurate:- Excellent low-end sensitivity
- Correlation with gold standard methods
- Precise throughput standard curve assay range
- Lot-to-lot consistency
Specifications
2°C to 8°C | |
Mycophenolic Acid | |
Control |
CEDIA Mycophenolic Acid Control 1 | |
Plasma | |
4 x 5 mL each level, low and high |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.