Learn More
Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassays, Calibrators and Controls
Complete your ISD panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay.
£301.00 - £4960.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Detectable Analytes | Mycophenolic Acid |
Sample Type | Plasma |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
17332743
|
Thermo Scientific™
100276 |
CEDIA Mycophenolic Acid Assay |
£4960.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17342743
|
Thermo Scientific™
100277 |
CEDIA Mycophenolic Acid Calibrator Set |
£301.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17352743
|
Thermo Scientific™
100278 |
CEDIA Mycophenolic Acid Control 1 |
£311.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17362743
|
Thermo Scientific™
100279 |
CEDIA Mycophenolic Acid Control 2 |
£311.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17372743
|
Thermo Scientific™
100280 |
CEDIA Mycophenolic Acid Control 3 |
£311.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complete your immunosuppressant drug testing panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay. Like our other ISD assays, the CEDIA Mycophenolic Acid Immunoassay is accurate and precise, providing users with confidence in their results. Applications are available on our flexible and intuitive Indiko™ and Indiko Plus clinical and specialty chemical systems as well as a wide variety of other laboratory instrumentation.
Key Features- Optimal low-end sensitivity with a broad dynamic range
- Excellent correlation with HPLC-MS, the gold standard method
- Precise throughout the standard curve assay range
Specifications
2°C to 8°C | |
Mycophenolic Acid | |
Plasma |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.