Learn More
Invitrogen™ eBioscience™ Cell Proliferation Dye eFluor™ 450
A violet fluorescent dye that can be used to monitor individual cell divisions for up to 7 generations.
Brand: Invitrogen™ 65-0842-85
92.00 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
- A violet fluorescent dye that can be used to monitor individual cell divisions
- Dye binds to any cellular proteins containing primary amines, and as cells divide, the dye is distributed equally between daughter cells
- They can be measured as successive halving of the fluorescence intensity of the dye - visualize up to 7 generations
- Cells labeled with Cell Proliferation Dye eFluor™ 450 may be fixed and permeabilized for analysis of intracellular targets using standard formaldehyde-containing fixatives and saponin-based permeabilization buffers
- Cell Proliferation Dye eFluor™ 450 has a peak excitation of 409 nm and can be detected using a 450/50 band pass filter (equivalent to eFluor™ 450 or Pacific Blue™), making it compatible with applications that utilize GFP
Additional Details
- Each vial may be reconstituted to a stock concentration of 10 mM with 165 μL of anhydrous DMSO
- Once reconstituted it should be used within 6 months and protected from light and stored at -20°C with dessicant
- Avoid freeze-thawing

Specifications
Violet | |
Proliferation Assay | |
Dye | |
Lyophilized powder | |
500 μg |
Fluorescence | |
Flow Cytometer | |
405/450 | |
eBioscience™ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.