missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Cell Proliferation Dye eFluor™ 450
A violet fluorescent dye that can be used to monitor individual cell divisions for up to 7 generations.
£138.00 - £256.00
Specifications
Color | Violet |
---|---|
Detection Method | Fluorescence |
For Use With (Application) | Proliferation Assay |
For Use With (Equipment) | Flow Cytometer |
Product Type | Dye |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15510597
|
Invitrogen™
65-0842-85 |
500 μg |
£138.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15520597
|
Invitrogen™
65-0842-90 |
4 x 500 μg |
£256.00
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- A violet fluorescent dye that can be used to monitor individual cell divisions
- Dye binds to any cellular proteins containing primary amines, and as cells divide, the dye is distributed equally between daughter cells
- They can be measured as successive halving of the fluorescence intensity of the dye - visualize up to 7 generations
- Cells labeled with Cell Proliferation Dye eFluor™ 450 may be fixed and permeabilized for analysis of intracellular targets using standard formaldehyde-containing fixatives and saponin-based permeabilization buffers
- Cell Proliferation Dye eFluor™ 450 has a peak excitation of 409 nm and can be detected using a 450/50 band pass filter (equivalent to eFluor™ 450 or Pacific Blue™), making it compatible with applications that utilize GFP
Additional Details
- Each vial may be reconstituted to a stock concentration of 10 mM with 165 μL of anhydrous DMSO
- Once reconstituted it should be used within 6 months and protected from light and stored at -20°C with dessicant
- Avoid freeze-thawing
Specifications
Violet | |
Proliferation Assay | |
Dye | |
Lyophilized powder |
Fluorescence | |
Flow Cytometer | |
405/450 | |
eBioscience™ |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ eBioscience™ Cell Proliferation Dye eFluor™ 450