Learn More
Invitrogen™ eBioscience™ Cell Stimulation Cocktail (500X)
Cocktail of phorbol 12-myristate 13-acetate (PMA) and ionomycin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines.
£70.75 - £203.00
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
15452024
|
Invitrogen™
00-4970-93 |
100uL |
£70.75
100µL |
In Stock
Log in to see stock. |
|||||
15290035
|
Invitrogen™
00-4970-03 |
4 x 100uL |
£203.00
400µL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Can be used to induce cytokine production in in vitro cell culture for subsequent detection in immunoassays
- Can also be used in conjunction with the Protein Transport Inhibitor Cocktail (500X) (cat. 00-4980) for intracellular retention of the induced cytokine expression
Specifications
eBioscience™ | |
Cell Stimulation Cocktail |
For Research Use Only. Not for use in diagnostic procedures.