Learn More
Molecular Biology Grade Water Tested to USP Sterile water, Corning™

Description
Molecular biology grade water is ideal for applications where highly pure, RNase-, DNase-, and protease-free water is required (e.g., PCR and other molecular biology reactions setup, DNA/RNA/protein extraction, purification, and storage reagents). Molecular biology grade water is tested to the chemical and physical properties found in the USP monograph for sterile purified water.
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility

Specifications
Specifications
Quantity | 6 x 1 L |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
24024018 | Certificate of Quality | 13/04/2025 | 46-000-CM |
18724009 | Certificate of Quality | 02/04/2025 | 46-000-CM |
24024017 | Certificate of Quality | 07/02/2025 | 46-000-CM |
17124007 | Certificate of Quality | 18/01/2025 | 46-000-CM |
14324001 | Certificate of Quality | 15/11/2024 | 46-000-CM |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.