missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ cellgro™ Ham's F-12

Brand: Corning™ 10-080-CM
This item is not returnable.
View return policy
Description
To assess growth promotion and cytotoxicity characteristics, Corning Classical Media products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Product performance is measured by harvest-to-plant ratios or fold increase according to predetermined specifications. Liquid and powder classical media products are also tested to determine pH and osmolality. Classical media powders are also tested for residual moisture. The pH and osmolality tests are performed according to specific procedures in which all equipment is calibrated using standards traceable to the National Institute of Standards and Technology. Biological testing of Corning liquid classical media products guarantees low levels of endotoxin, mycoplasma, and an absence of contamination. Endotoxins: classical media products are tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay. For release, each lot of standard liquid classical media must have an endotoxin level of less than 0.25 EU/mL. Classical media powder products must have an endotoxin level of less than 0.25 EU/mL when reconstituted to formula weight with low endotoxin water. All Corning liquid classical media products are tested for sterility and liquid classical media products are tested for mycoplasma off-site using a large-volume method.Order Info
dimQuantity

Specifications
2 to 8°C | |
Ham's F-12 | |
Inorganic Salts: 33.3mg/L CaCl2 (anhydrous), 0.0016mg/L CuSO2 (anhydrous), 0.834mg/L FeSO4.7H2O, 223.6mg/L KCl, 72.20mg/L MgSO4 (anhydrous), 7600mg/L NaCl, 142mg/L Na2HPO4 (anhydrous), 1176mg/L NaHCO3, 0.863mg/L ZnSO4.7H2O; Amino Acids: 8.9mg/L L-Alanine, 211mg/L L-Arginine. HCl, 15mg/L L-Asparagine.H2O, 13mg/L L-Aspartic acid, 35.12mg/L L-Cysteine.HCl.H2O, 14.70mg/L L-Glutamic acid, 146.20mg/L L-glutamine, 7.50mg/L Glycine, 20.96mg/L L-Histidine.HCl.H2O, 3.94mg/L L-Isoleucine, 13.10mg/L L-Leucine, 36.50mg/L L-Lysine. HCl, 4.48mg/L L-Methionine, 4.96mg/L L-Phenylalanine, 34.50mg/L L-Proline, 10.50mg/L L-Serine, 11.90mg/L L-Threonine, 2.04mg/L L-Tryptophan, 7.84mg/L L-Tyrosine.2Na.2H2O, 11.70mg/L L-Valine; Vitamins: 0.037mg/L Nicotinamide, 0.062mg/L Pyridoxine. HCl, 0.038mg/L Riboflavin, 0.34mg/L Thiamine. HCl, 1.36mg/L Vitamin B12; Other: 1802mg/L D-Glucose, 4.77mg/L Hypoxanthine, Na, 0.21mg/L DL-Thioctic (lipoic) acid, 0.088mg/L Methyl lineoleate, 1.2mg/L Phenol red. Na, 0.16mg/L Putrescine.2HCl, 110mg/L Sodium pyruvate, 0.73mg/L Thymidine. | |
6/Pk. |
1 X | |
<0.25EU/mL | |
To assess growth promotion and cytotoxicity characteristics |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ cellgro™ Ham's F-12 > Capacity: 1L
Spot an opportunity for improvement?Share a Content Correction